Learn More
MilliporeSigma™ Perforated Silicone Stoppers
Catalog No. XX1002508 Shop All MilliporeSigma Products
Change view
Click to view available options
Stopper Number:
5
Catalog No. Stopper Number
XX1002508 5
1 options
Catalog No. XX1002508 Supplier MilliporeSigma™ Supplier No. XX1002508
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Perforated Stopper
Stopper Number 5
Size 5
Material Silicone
For Use With (Equipment) 25mm Millipore™ vacuum filtration flasks and filter holder