Learn More
MilliporeSigma™ pET-11a DNA Vector
Catalog No. 694363 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-436-3 10 μg
1 options
Catalog No. 69-436-3 Supplier MilliporeSigma™ Supplier No. 694363
Add to Cart
Add to Cart
The pET-11a-d vectors carry an N-terminal T7·Tag™ sequence and BamH I cloning site. These vectors are the precursors to many pET family vectors; sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Restriction Site BamH I
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg