Learn More
MilliporeSigma™ pET-14b DNA Vector
Catalog No. 696603 Shop All MilliporeSigma Products
Change view
Click to view available options
Content And Storage:
Store at -70°C. Avoid freeze/thaw cycles.
Catalog No. Content And Storage
69-660-3 Store at -70°C. Avoid freeze/thaw cycles.
1 options
Catalog No. 69-660-3 Supplier MilliporeSigma™ Supplier No. 696603
Add to Cart
Add to Cart
The pET-14b vector carries an N-terminal His·Tag™ sequence followed by a thrombin cleavage site and three cloning sites. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.