Learn More
MilliporeSigma™ pET-15b DNA Vector
Catalog No. 696613 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-661-3 10 μg
1 options
Catalog No. 69-661-3 Supplier MilliporeSigma™ Supplier No. 696613
Add to Cart
Add to Cart
The pET-15b vector carries an N-terminal His·Tag™ sequence followed by a thrombin cleavage site and three cloning sites. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg