Learn More
MilliporeSigma™ pET-16b DNA Vector
Catalog No. 696623 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-662-3 10 μg
1 options
Catalog No. 69-662-3 Supplier MilliporeSigma™ Supplier No. 696623
Add to Cart
Add to Cart
The pET-16b vector carries an N-terminal His·Tag™ sequence followed by a Factor Xa site and three cloning sites. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at - 70°C. Avoid freeze/thaw cycles.
Quantity 10 μg