Learn More
MilliporeSigma™ pET-17b DNA Vector
Catalog No. 696633 Shop All MilliporeSigma Products
Change view
Click to view available options
Restriction Site:
Dual BstX I sites
Catalog No. Restriction Site
69-663-3 Dual BstX I sites
1 options
Catalog No. 69-663-3 Supplier MilliporeSigma™ Supplier No. 696633
Add to Cart
Add to Cart
The pET-17b vector carries an N-terminal 11aa T7·Tag™ sequence followed by a region of useful cloning sites. Included in the multiple cloning region are dual BstX I sites, which allow efficient cloning using an asymmetric linker (1). Unique sites (except for the two BstX I sites) are shown on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Restriction Site Dual BstX I sites
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.