Learn More
MilliporeSigma™ pET-19b DNA Vector
Catalog No. 696773 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-677-3 10 μg
1 options
Catalog No. 69-677-3 Supplier MilliporeSigma™ Supplier No. 696773
Add to Cart
Add to Cart
The pET-19b vector carries an N-terminal His·Tag™ sequence followed by an enterokinase cleavage site and three cloning sites. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg