Learn More
MilliporeSigma™ pET-20b(+) DNA Vector
Catalog No. 697393 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-739-3 10 μg
1 options
Catalog No. 69-739-3 Supplier MilliporeSigma™ Supplier No. 697393
Add to Cart
Add to Cart
The pET-20b(+) vector carries an N-terminal pelB signal sequence for potential periplasmic localization, plus optional C-terminal His·Tag™ sequence. Unique sites are shown on the vector map; note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg