Learn More
MilliporeSigma™ pET-23(+) DNA Vector
Catalog No. 697713 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-77-13 10 μg
1 options
Catalog No. 69-77-13 Supplier MilliporeSigma™ Supplier No. 697713
Add to Cart
Add to Cart
The pET-23(+) is transcription vector designed for expression from bacterial translation signals carried within a cloned insert. It therefore lacks the ribosome binding site and ATG start codon present on the pET translation vectors. A C-terminal His·Tag™ sequence is available. The sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg