Learn More
MilliporeSigma™ pET-23b(+) DNA Vector
Catalog No. 697463 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-746-3 10 μg
1 options
Catalog No. 69-746-3 Supplier MilliporeSigma™ Supplier No. 697463
Add to Cart
Add to Cart
The pET-23a-d(+) vectors carry an N-terminal T7·Tag™ sequence plus an optional C-terminal His·Tag™ sequence. The sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map. The f1 origin is oriented so that infection with helper phage will produce virions containing single-stranded DNA that corresponds to the coding strand.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg