Learn More
MilliporeSigma™ pET-26b(+) DNA Vector
Catalog No. 698623 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-862-3 10 μg
1 options
Catalog No. 69-862-3 Supplier MilliporeSigma™ Supplier No. 698623
Add to Cart
Add to Cart
The pET-26b(+) vector carries an N-terminal pelB signal sequence for potential periplasmic localization, plus optional C-terminal His·Tag™ sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg