Learn More
MilliporeSigma™ pET-28a(+) DNA Vector
GSA_VA
Catalog No. 698643 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-864-3 10 μg
1 options
Catalog No. 69-864-3 Supplier MilliporeSigma™ Supplier No. 698643
Add to Cart
Add to Cart
The pET-28a-c(+) vectors carry an N-terminal His·Tag™/thrombin/T7·Tag™ configuration plus an optional C-terminal His·Tag sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg