Learn More
MilliporeSigma™ pET-28b(+) DNA Vector
Catalog No. 698653 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-865-3 10 μg
1 options
Catalog No. 69-865-3 Supplier MilliporeSigma™ Supplier No. 698653
Add to Cart
Add to Cart
The pET-28a-c(+) vectors carry an N-terminal His·Tag™/thrombin/T7·Tag™ configuration plus an optional C-terminal His·Tag sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg