Learn More
MilliporeSigma™ pET-30a(+) DNA Vector
Catalog No. 699093 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-909-3 10 μg
1 options
Catalog No. 69-909-3 Supplier MilliporeSigma™ Supplier No. 699093
Add to Cart
Add to Cart
The pET-30a-c(+) vectors carry an N-terminal His·Tag™/thrombin/S·Tag™/enterokinase configuration plus an optional C-terminal His·Tag sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycle.
Quantity 10 μg