Learn More
MilliporeSigma™ pET-30c(+) DNA Vector
Catalog No. 699113 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-911-3 10 μg
1 options
Catalog No. 69-911-3 Supplier MilliporeSigma™ Supplier No. 699113
Add to Cart
Add to Cart
The pET-30a-c(+) vectors carry an N-terminal His·Tag™/thrombin/S·Tag™/enterokinase configuration plus an optional C-terminal His·Tag sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store mat -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg