Learn More
MilliporeSigma™ pET-31b(+) DNA Vector
Catalog No. 699523 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
69-952-3 10 μg
1 options
Catalog No. 69-952-3 Supplier MilliporeSigma™ Supplier No. 699523
Add to Cart
Add to Cart
The pET-31b(+) vector is designed for cloning and high-level expression of peptide sequences fused with the 125aa ketosteroid isomerase protein (1). The unique AlwN I cloning site allows the unidirectional insertion of of tandemly repeated peptide coding regions separated by methionine codons. The sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg