Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pET-39b(+) DNA Vector

Catalog No. 700903 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-090-3 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-090-3 Supplier MilliporeSigma™ Supplier No. 700903
Add to Cart
Add to Cart
The pET-39b(+) vector is designed for expression of DsbA fusion proteins. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg
For Use With (Application) Expression of DsbA fusion proteins

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.