Learn More
MilliporeSigma™ pET-40b(+) DNA Vector
Catalog No. 700913 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-091-3 10 μg
1 options
Catalog No. 70-091-3 Supplier MilliporeSigma™ Supplier No. 700913
Add to Cart
Add to Cart
The pET-39b(+) vector is designed for expression of DsbA fusion proteins. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg