Learn More
MilliporeSigma™ pET-41 Ek/LIC Vector Kit
Catalog No. 710713 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
71-071-3 20 Reactions
1 options
Catalog No. 71-071-3 Supplier MilliporeSigma™ Supplier No. 710713
Add to Cart
Add to Cart
The pET-41 series is designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST·Tag™ protein. Vector encoded sequence can be completely removed by cleaving the GST fusion protein with enterokinase.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -70°C.
Quantity 20 Reactions