Learn More
MilliporeSigma™ pET-41a(+) DNA Vector
Catalog No. 705563 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-556-3 10 μg
1 options
Catalog No. 70-556-3 Supplier MilliporeSigma™ Supplier No. 705563
Add to Cart
Add to Cart
The pET-41 series is designed for cloning and high-level expression of peptide sequences fused with the 220 aa GST·Tag™ protein. The sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. Vector encoded sequence can be completely removed when cloning into the PshAI site (as shown below) and then cleaving the GST fusion protein with Enterokinase.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 10 μg