Learn More
MilliporeSigma™ pET-42b(+) DNA Vector
Catalog No. 705623 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-562-3 10 μg
1 options
Catalog No. 70-562-3 Supplier MilliporeSigma™ Supplier No. 705623
Add to Cart
Add to Cart
The pET -42 series feature the powerful T7lac promoter, and encode the GST·Tag (220 aa) sequence, proteolytic sites, His·Tag (6 aa) sequence, and S·Tag (15 aa) sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at - 70°C. Avoid freeze/thaw cycles.
Quantity 10 μg