Learn More
MilliporeSigma™ pET-43.1a(+) DNA Vector
Catalog No. 709393 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-939-3 10 μg
1 options
Catalog No. 70-939-3 Supplier MilliporeSigma™ Supplier No. 709393
Add to Cart
Add to Cart
The pET-43.1 series of vectors are designed for cloning and high-level expression of peptide sequences fused with the 491 aa Nus·Tag™ protein, the sequence is numbered by the pBR322 convention so the T7 expression region is reversed on the vector map, TB288VM. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. Vector encoded sequence can be completely removed when cloning into the PshA I or Sma I sites by cleaving the Nus·Tag fusion protein with Enterokinase or Thrombin, respectively.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -70°C.
Quantity 10 μg