Learn More
MilliporeSigma™ pET-44 Ek/LIC Vector Kit
Catalog No. 711443 Shop All MilliporeSigma Products
Change view
Click to view available options
pET-44 Ek/LIC Vector Kit
Catalog No.
71-144-3 pET-44 Ek/LIC Vector Kit
1 options
Catalog No. 71-144-3 Supplier MilliporeSigma™ Supplier No. 711443/DEL/DEL
The pET-44 vectors incorporate the Nus·Tag™ technology, which allows target proteins to be expressed as fusions to the highly soluble NusA protein. According to a modeling study, the NusA protein has the highest potential for solubility among more than 4000 E. coli proteins, and therefore increases the proportion of soluble protein when expressed in E. coli as a fusion partner. Ek/LIC represents a highly flexible cloning strategy in which a single insert preparation can be simultaneously cloned into a wide variety of compatible Ligation Independent Cloning (LIC) vectors to allow high-throughput optimization of target protein expression.