Learn More
MilliporeSigma™ pET-45b(+) DNA Vector
Catalog No. 713273 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
71-327-3 10 μg
1 options
Catalog No. 71-327-3 Supplier MilliporeSigma™ Supplier No. 713273
Add to Cart
Add to Cart
The pET-45b plasmid features the ability to express fusion proteins with an N-terminal His·Tag™ coding sequence that results in native protein after purification and cleavage. The plasmid contains a strong T7lac promoter, an amino-terminal His·Tag coding sequence, and multiple cloning site (MCS) regions designed to allow the generation of target proteins with minimal vector-encoded fusion. The Pml I cloning site allows direct fusion to the His·Tag sequence for inserts that are blunt and in the appropriate reading frame.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7lac
Content And Storage Store at or below -70°C.
Quantity 10 μg