Learn More
MilliporeSigma™ pET-47b(+) DNA Vector
Catalog No. 714613 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
71-461-3 10 μg
1 options
Catalog No. 71-461-3 Supplier MilliporeSigma™ Supplier No. 714613
Add to Cart
Add to Cart
The pET-47b(+) plasmid features the ability to express fusion proteins with an N-terminal His·Tag™ coding sequence that is cleavable with the human rhinovirus (HRV) 3C protease. The plasmid contains a strong T7lac promoter, an optimized RBS, the coding sequence for the HRV 3C protease cleavage site (LeuGluValLeuPheGlnGlyPro), and a multiple cloning site that contains restriction enzyme sites found in many other Novagen expression vectors to facilitate insert transfer. An optional C-terminal S·Tag™ coding sequence is compatible with purification, detection and quantification.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7lac
Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) Express fusion proteins with an N-terminal HisTag™ coding sequence that is cleavable with the human rhinovirus (HRV) 3C protease