Learn More
MilliporeSigma™ pET-49b(+) DNA Vector
Catalog No. 714633 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
71-463-3 10 μg
1 options
Catalog No. 71-463-3 Supplier MilliporeSigma™ Supplier No. 714633
Add to Cart
Add to Cart
The pET-49b(+) vector is designed for cloning and high-level expression of target proteins fused with the 220 aa schistosomal glutathione-S-transferase (GSTTag™) protein that is cleavable with the human rhinovirus (HRV) 3C protease.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -70°C.
Quantity 10 μg