Learn More
MilliporeSigma™ pET-51 Ek/LIC Vector Kit
Catalog No. 715703 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
71-570-3 20 Reactions
1 options
Catalog No. 71-570-3 Supplier MilliporeSigma™ Supplier No. 715703
Add to Cart
Add to Cart
The pET-51Ek/LIC Vector is an Ek/LIC version of pET-51b(+). It is designed for cloning and high-level expression of target proteins fused with the 8 aa Strep·Tag™ II coding sequence that is cleavable with enterokinase (Ek) protease. The plasmid contains a strong T7lac promoter, an optimized RBS, the coding sequence for the Ek protease cleavage site (AspAspAspAspLys(down arrow)), and a multiple cloning site that contains restriction enzyme sites found in many other Novagen expression vectors to facilitate insert transfer.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7lac
Content And Storage Store at or below -70°C.
Quantity 20 Reactions
For Use With (Application) Cloning and high-level expression of target proteins fused with the 8 aa StrepTag™ II coding sequence that is cleavable with enterokinase (Ek) protease