Learn More
MilliporeSigma™ pET-52b(+) DNA Vector
Catalog No. 715543 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
71-554-3 10 μg
1 options
Catalog No. 71-554-3 Supplier MilliporeSigma™ Supplier No. 715543
Add to Cart
Add to Cart
The pET-52b(+) vector is designed for cloning and high-level expression of target proteins fused with the 8 aa Strep·Tag™ II coding sequence that is cleavable with human rhinovirus (HRV) 3C protease. The plasmid contains a strong T7lac promoter, an optimized RBS, the coding sequence for the HRV 3C protease cleavage site (LysGluValLysPheGl(down arrow)GlyPro), and a multiple cloning site that contains restriction enzyme sites found in many other Novagen expression vectors to facilitate insert transfer. It also encodes an optional C-terminal thrombin protease cleavage site and His·Tag™ coding sequence with 10 histidines.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7lac
Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) Cloning and high-level expression of target proteins fused with the 8 aa StrepTag™ II coding sequence that is cleavable with human rhinovirus (HRV) 3C protease