Learn More
MilliporeSigma™ pET Expression System 30
Catalog No. 707813 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
Up to 10 transformations
Catalog No. Quantity
70-781-3 Up to 10 transformations
1 options
Catalog No. 70-781-3 Supplier MilliporeSigma™ Supplier No. 707813
Add to Cart
Add to Cart
The pET Expression System 3 contains 10μg each of the four versions of pET-30 (pET-30a-c(+)). The pET-30a-c(+) vectors carry an N-terminal His·Tag™/thrombin/S·Tag™/enterokinase configuration plus an optional C-terminal His·Tag sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map, TB095VM.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity Up to 10 transformations