Learn More
MilliporeSigma™ pETBlue-2 Uncut DNA Vector
Catalog No. 706093 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
70-609-3 20 μg
1 options
Catalog No. 70-609-3 Supplier MilliporeSigma™ Supplier No. 706093
Add to Cart
Add to Cart
The pETBlue™-2 vector is designed to identify recombinants by traditional blue/white screening while also allowing T7lac promoter based expression of target genes. Screening is independent of expression because the T7lac expression promoter is in an opposed orientation relative to the E. coli promoter that mediates blue/white screening. pETBlue-2 defines the open reading frame and inserts must be cloned in-frame if expression is desired.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 20 μg