Learn More
MilliporeSigma™ pETcoco-1 DNA Expression Vector
Catalog No. 711293 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
71-129-3 10 μg
1 options
Catalog No. 71-129-3 Supplier MilliporeSigma™ Supplier No. 711293
Add to Cart
Add to Cart
The pETcoco™-1 expression vector is designed to allow on command amplification of vector copy number from single copy (1–2 plasmid copies per cell) to medium copy (20–50 per cell). The vector is maintained in the single copy state by utiliziing the oriS and repE elements of the F episome together with the parABC partition determinants. The medium copy state is obtained by employing the RK-2 derived trfA replicator acting at oriV.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -70°C.
Quantity 10 μg