Learn More
MilliporeSigma™ pETcoco-2 Expression System
Catalog No. 711493 Shop All MilliporeSigma Products
Change view
Click to view available options
Content And Storage:
Store at or below -70°C.
Catalog No. Content And Storage
71-149-3 Store at or below -70°C.
1 options
Catalog No. 71-149-3 Supplier MilliporeSigma™ Supplier No. 711493
Add to Cart
Add to Cart
The pETcoco™ System combines the advantages of T7 promoter-driven protein expression with the ability to control plasmid copy number (Sektas 2003). In the presence of 0.2% glucose, pETcoco recombinant plasmids are maintained at one copy per cell. In this single-copy state, pETcoco clones are extremely stable because background expression and consequent gene rearrangements or deletions are virtually absent.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7
Content And Storage Store at or below -70°C.