Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pETDuet-1 DNA Vector
GSA_VA

Catalog No. 711463 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-146-3 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-146-3 Supplier MilliporeSigma™ Supplier No. 711463
Add to Cart
Add to Cart
pETDuet™-1 is designed for the coexpression of two target genes. The vector encodes two multiple cloning sites (MCS), each of which is preceded by a T7 promoter, lac operator and ribosome binding site (rbs). The vector also carries the pBR322-derived ColE1 replicon, lacI gene and ampicillin resistance gene. This vector can be transformed into the same cell with pACYCDuet™-1, pCDFDuet™-1, and pRSFDuet™-1 or pCOLADuet™-1 in the appropriate host strain for the coexpression of up to eight target genes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) Coexpression of two target genes

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.