Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Petri Dishes and Petri-Pad™ Dishes
GSA_VA

Catalog No. pd2004705 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Dish with Pad
Dish without Pad
Quantity:
100 Pk. (5 Sleeves)
150 Pack
500 Pk. (25 Sleeves)
600 Pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
PD2004705 Dish without Pad 600 Pack
PD20047S5 Dish with Pad 500 Pk. (25 Sleeves)
PD20047S0 Dish with Pad 150 Pack
PD2004700 Dish without Pad 100 Pk. (5 Sleeves)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PD2004705 Supplier MilliporeSigma™ Supplier No. PD2004705
Only null left
Add to Cart
Add to Cart

Requires

When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 045 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)

Available with or without absorbent pads

Petri Dishes

  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Single-use
Material Polystyrene
Shape Round
Sterility Sterile
Type Dish without Pad
For Use With (Application) For solid agar medium or liquid medium with sterile absorbent pads
Packaging Sleeve
Quantity 600 Pack
Diameter (Metric) 47 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.