Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Phospho-CREB (Ser133), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710131MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-31MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-31MI Supplier MilliporeSigma™ Supplier No. 1710131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Phospho-CREB (Ser133) Clone: in Hamster, Human, Mouse, Rat samples, and it is validated for ChIP, Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blotting

CREB is a β-ZIP transcription factor that activates target genes through cAMP response elements. CREB is able to mediate signals from numerous physiological stimuli, resulting in regulation of a broad array of cellular responses. While CREB is expressed in numerous tissues, it plays a large regulatory role in the nervous system. CREB is believed to play a key role in promoting neuronal survival, precursor proliferation, neurite outgrowth and neuronal differentiation in certain neuronal populations. CREB promotes outgrowth and differentiation as a mediator of neurotrophin pathways. CREB is able to selectively activate numerous downstream genes through interactions with different dimerization partners. CREB is activated by phosphorylation at Ser133 by various signaling pathways including Erk, Ca2+ and stress signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CREB (Ser133)
Applications ChIP Assay, Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-phospho-CREB (Ser133) (rabbit polyclonal). One vial containing 100μg of affinity purified rabbit serum in 0.1M Tris-Glycine (pH 7.4) 0.15M NaCl, and 0.05% sodium azide, before the addition of 30% glycerol. Normal Rabbit IgG. One vial containing 125μg Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers cFos CRE. One vial containing 75μL of 5μM of each primer specific for the cFos CRE. FOR: GGC CCA CGA GAC CTC TGA GAC A; REV: GCC TTG GCG CGT GTC CTA ATC T
Gene Accession No. P16220
Gene Symbols CREB1, CREB, MGC9284
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to the amino acids including and encompassing phosphorylated Ser133 of CREB.
Purification Method Purified by protein A-Sepharose followed by immunoadsorbant chromatography using an unphosphorylated CREB affinity gel column
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Signaling, Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_134442
Test Specificity Recognizes p43 phosphorylated CREB. May also recognize p30 and p38kDa proteins. The p30 & p38 proteins may be phosphorylated CREM and ATF-1 respectively, since the two proteins share significant homology to the phosphorylated CREB immunizing peptide.
Includes This ChIPAb+ Phospho-CREB (Ser133) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Hamster, Human, Mouse, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.