Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Phospho-Histone H3 (Ser28), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710269MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-69MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-69MI Supplier MilliporeSigma™ Supplier No. 1710269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Phospho-Histone H3 (Ser28) Clone: in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Immunocytochemistry, Peptide Inhibition Assay, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Histone H3 (Ser28)
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti- Phospho-Histone H3 (Ser28) (Rabbit Polyclonal). One vial containing 125μl of purified rabbit polyclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide before the addition of 30% glycerol. Normal Rabbit IgG. Two vials containing 75μL of normal rabbit IgG in buffer containing 0.01M PBS with 0.1% sodium azide. ChIP Primers, human GAPDH promoter. One vial containing 75μL of 5μM each primer specific for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. Q16695
Gene Symbols HIST3H3, H3FT
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to Histone H3 phosphorylated at Ser28.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Includes This ChIPAb+ Phospho-Histone H3 (Ser28) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.