Learn More
MilliporeSigma™ pIE1-neo DNA Vector
Catalog No. 701713 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-171-3 10 μg
1 options
Catalog No. 70-171-3 Supplier MilliporeSigma™ Supplier No. 701713
Add to Cart
Add to Cart
The pIE1-neo plasmid is designed for the establishment of stably-transformed insect cells. When co-transfected with a pIE1 vector containing a foreign gene under the control of the baculovirus ie1 promoter, stable cell lines that constitutively express low levels of the target gene can be selected in the presence of the antibiotic G418 (1).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 10 μg