Learn More
MilliporeSigma™ pIEx-10 DNA Vector
Catalog No. 715573 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Application):
Cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells
Catalog No. For Use With (Application)
71-557-3 Cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells
1 options
Catalog No. 71-557-3 Supplier MilliporeSigma™ Supplier No. 715573
Add to Cart
Add to Cart
The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. The pIEx-10 contains the coding sequence for the mouse IgM signal sequence to allow secretion of the expressed protein. It also encodes an N-terminal 8 aa Strep·Tag™ II coding sequence followed by an enterokinase protease cleavage site and a multiple cloning site, with the option to include a C-terminal His·Tag™ coding sequence with 10 histidines.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells