Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pIEx-10 Ek/LIC Vector Kit

Catalog No. 715743 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-574-3 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-574-3 Supplier MilliporeSigma™ Supplier No. 715743
Add to Cart
Add to Cart
The pIEx™-10 Ek/LIC vector is an Ek/LIC version of pIEx-10. The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. The pIEx-10 Ek/LIC vector contains the coding sequence for the mouse IgM signal sequence to allow secretion of the expressed protein. It also encodes an N-terminal 8 aa Strep·Tag™ II coding sequence followed by an enterokinase protease cleavage site and a multiple cloning site, with the option to include a C-terminal His·Tag™ coding sequence with 10 histidines.
TRUSTED_SUSTAINABILITY

Specifications

Promoter IE1
Content And Storage Store at or below -70°C.
Quantity 20 Reactions
For Use With (Application) Cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.