Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pIEx-6 DNA Vector

Catalog No. 713333 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-333-3 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-333-3 Supplier MilliporeSigma™ Supplier No. 713333
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The pIEx™-6 vector is designed to allow rapid, high-level protein expression in insect cells without the time-consuming process of creating a recombinant baculovirus. This vector features the hr5 enhancer and the IE1 (immediate early) promoter to direct expression in insect cells. This promoter/enhancer combination recruits endogenous insect cell transcription machinery, thereby avoiding baculovirus infection and the associated cytotoxic effects. This plasmid features an amino-terminal His·Tag™ coding sequence, and multiple cloning site (MCS) regions designed to allow the generation of target proteins with minimal vector-encoded fusion.
TRUSTED_SUSTAINABILITY

Specifications

Promoter IE1
Content And Storage Store at or below -70°C.
Quantity 20 μg
For Use With (Application) Rapid, high-level protein expression in insect cells without creating a recombinant baculovirus

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.