Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pIEx/Bac-1 DNA vector

Catalog No. 717243 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-724-3 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-724-3 Supplier MilliporeSigma™ Supplier No. 717243
Add to Cart
Add to Cart
The pIEx/Bac™ vectors are dual-purpose vectors that offer unique flexibility for cloning and expression of proteins in Spodoptera-derived insect cells. These vectors contain the Autographa californica nuclear polyhedrosis virus (AcNPV) enhancer/immediate early promoter combination (hr5/ie1) for plasmid-mediated and early baculovirus expression, and the AcNPV p10 very late promoter for robust baculovirus-mediated expression. The pIEx/Bac-1 vector contains an N-terminal 8 aa Strep·Tag™ II coding sequence followed by an enterokinase protease cleavage site and a multiple cloning site, with the option to include a C-terminal His·Tag™ coding sequence with 10 histidines.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Quantity 20 μg
For Use With (Application) Cloning and expression of proteins in Spodoptera-derived insect cells

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.