Learn More
MilliporeSigma™ Pipe Thread Fittings
Catalog No. p-3303595 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Pipe Thread Fitting
Connection Type:
1/4 in. NPFT Coupling
1/4 in. NPTM
1/4 in. NPTM Close Nipple
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Connection Type
XX6700101 Pipe Thread Fitting 1/4 in. NPTM
XX6700115 Pipe Thread Fitting 1/4 in. NPTM Close Nipple
XX6700111 Pipe Thread Fitting 1/4 in. NPFT Coupling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. XX6700101 Supplier MilliporeSigma™ Supplier No. XX6700101
Only null left
Add to Cart
Add to Cart
Use in conjunction with filtration systems.
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type 1/4 in. NPTM
For Use With (Equipment) Water Purification System
Type Pipe Thread Fitting
Material Series 300 stainless steel
Diameter (English) Inside 0.25 in.
Diameter (Metric) Inside 0.63 cm