Learn More
MilliporeSigma™ PKA, Catalytic Subunit, Mouse, Recombinant Protein, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 50mM NaCl, 20mM Tris-HCl, 1mM EDTA, 2mM DTT, 50% glycerol, pH 7.5. |
| Quantity | 100 kU |
| Source | E. coli |
| Immunogen | PKA, Catalytic Subunit |
| Storage Requirements | Following initial thaw, aliquot and freeze (−20°C). |
| Regulatory Status | RUO |
| Species | Mouse |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.