Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Plasmin, Human Plasma, Protein

Catalog No. 52762110U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-762-110U 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-762-110U Supplier MilliporeSigma™ Supplier No. 52762110U
Add to Cart
Add to Cart
Native plasmin from human plasma. Catalyzes the hydrolysis of peptides, arginine and lysine esters and converts fibrin into soluble products.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM sodium phosphate, 1mM 6-aminohexanoic acid, 25% glycerol, pH 7.3.
Quantity 10 U
Immunogen Plasmin
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.