Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 20mM Tris-HCl, 2mM EDTA, pH 7.5. |
| Preparation Method | H2O |
| Purification Method | SDS-PAGE |
| Quantity | 120 U |
| Immunogen | Plasminogen |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 1 week at 4°C or for up to 2 months at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.