Learn More
MilliporeSigma™ pLysS DNA Vector
Catalog No. 696593 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 μg
Catalog No. Quantity
69-659-3 1 μg
1 options
Catalog No. 69-659-3 Supplier MilliporeSigma™ Supplier No. 696593
Add to Cart
Add to Cart
These plasmids are not cloning vectors; they are used in λDE3 lysogenic hosts to suppress basal expression from the T7 promoter by producing T7 lysozyme, a natural inhibitor of T7 RNA polymerase.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 1 μg