Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Polishing Cartridge

Catalog No. CDEXP0LUV Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Milli-Q™ UV water system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
CDEXP0LUV Milli-Q™ UV water system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDEXP0LUV Supplier MilliporeSigma™ Supplier No. CDEXP0LUV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Milli-Q

For Milli-Q™ UV system
TRUSTED_SUSTAINABILITY

Specifications

Purification Method Ion Exchange, Activated Carbon
For Use With (Equipment) Milli-Q™ UV water system
Type Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.