Learn More
MilliporeSigma™ Precoated Plastic-Backed TLC Sheets
Catalog No. M55817 Shop All MilliporeSigma Products
Catalog No. M55817 Supplier MilliporeSigma™ Supplier No. 55817
For analytical TLC analysis
  • Use of standardized and controlled sorbents
  • Sorbent: Aluminum Oxide 60, Neutral F254
  • Layer Thickness: 200μm