Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prekallikrein, Human Plasma, Protein

Catalog No. 5295831MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-958-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-958-31MG Supplier MilliporeSigma™ Supplier No. 5295831MG
Add to Cart
Add to Cart
Native prekallikrein from human plasma. Single-chain glycoprotein that participates in the early phase of blood coagulation, kinin formation, and fibrinolysis.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 150mM NaCl, 4mM NaOAc-HCl, pH 5.3.
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Prekallikrein
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.