Learn More
Specifications
Specifications
| Formulation | Liquid in 10mM sodium acetate buffer, pH 4.0, containing 0.01% sodium azide. |
| Gene ID (Entrez) | NM_000311.3, NM_001080121.1, NM_001080122.1, NM_001080123.1, NM_183079.2 |
| Name | Prion Protein, recombinant |
| Quantity | 50 μg |
| Storage Requirements | -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.